Gene Transcripts

Gene Symbol
Klk1b24
Entrez Gene ID
16617
Gene Name
kallikrein 1-related peptidase b24
Gene Aliases
Klk24, mGk-24
Chromosome Location
Chr. 7: 44188230 - 44192455 on Build GRCm38
UniGene ID
Mm.483098
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010643.1 NP_034773.1 338
GenBank mRNA AB039277.1 - 338
BC146015.1 - 3 365 40131318
BC146019.1 - 3 365 40131322

View Full Product Details