Gene Transcripts
- Gene Symbol
- Klra8
- Entrez Gene ID
- 16639
- Gene Name
- killer cell lectin-like receptor, subfamily A, member 8
- Gene Aliases
- Cmv-1, Cmv1, Ly49h, Ly49u<129>
- Chromosome Location
- Chr. 6: 130115226 - 130129898 on Build GRCm38
- UniGene ID
- Mm.439698
- Species
- Mus musculus
- Gene Symbol
- Klra14-ps
- Entrez Gene ID
- 654449
- Gene Name
- killer cell lectin-like receptor subfamily A, member 14, pseudogene
- Gene Aliases
- EG654449, Gm15858, Klra14, Klra29, Klra31, Ly49N, OTTMUSG00000026959
- Chromosome Location
- Chr. 6: 130149106 - 130160748 on Build GRCm38
- UniGene ID
- Mm.439698
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY860974.1 |
- |
1, 2 |
87 |
|
- Gene Symbol
- Klra21
- Entrez Gene ID
- 93968
- Gene Name
- killer cell lectin-like receptor subfamily A, member 21
- Gene Aliases
- Klra-ps1, Ly49H, Ly49u
- Chromosome Location
- Chr. 6: 130082598 - 130096486 on Build GRCm38
- UniGene ID
- Mm.439698
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC116399.1 |
- |
|
197 |
|
View Full Product Details