Gene Transcripts
- Gene Symbol
- Klrc2
- Entrez Gene ID
- 16642
- Gene Name
- killer cell lectin-like receptor subfamily C, member 2
- Gene Aliases
- NKG2C
- Chromosome Location
- Chr. 6: 129649602 - 129660737 on Build GRCm38
- UniGene ID
- Mm.347927
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF106010.1 |
- |
6, 7 |
883 |
|
View Full Product Details