Gene Transcripts

Gene Symbol
Ccndbp1
Entrez Gene ID
17151
Gene Name
cyclin D-type binding-protein 1
Gene Aliases
AU022347, DIP1, GCIP, Maid, SECC-8, SSEC-8
Chromosome Location
Chr. 2: 121008383 - 121016912 on Build GRCm38
UniGene ID
Mm.7838
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010761.2 NP_034891.2 4, 5 431
GenBank mRNA AK144883.1 - 4, 5 408
AK154285.1 - 4, 5 385
AK160205.1 - 4, 5 397
AK163150.1 - 4, 5 344
AK167345.1 - 4, 5 396
AK186314.1 - 3, 4 208
BC001984.1 - 4, 5 367 3489973
BC005802.1 - 3, 4 440 3707919
CO810054.1 - 4, 5 438
U50734.1 - 4, 5 349

View Full Product Details