Gene Transcripts

Gene Symbol
Pik3c2g
Entrez Gene ID
18705
Gene Name
phosphatidylinositol 3-kinase, C2 domain containing, gamma polypeptide
Gene Aliases
C80387, PI3K-C2-gamma
Chromosome Location
Chr. 6: 139614072 - 139657235 on Build GRCm38
UniGene ID
Mm.333471
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011084.2 NP_035214.2 2 368
NM_207683.2 NP_997566.1 3114
GenBank mRNA AB008791.1 - 3114
AB008792.1 - 2 363
AK036916.1 - 2 179
AK054266.1 - 2 159
AK054271.1 - 2 244
AK054415.1 - 2 311
AK076317.1 - 2769
BC061142.1 - 2 155
BC150813.1 - 3106 9087724

View Full Product Details