Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001253357.1 | NP_001240286.1 | 3 | 517 | |
NM_005424.4 | NP_005415.1 | 3 | 600 | ||
XM_005271163.1 | XP_005271220.1 | 3 | 543 | ||
XM_006710869.1 | XP_006710932.1 | 3 | 734 | ||
XM_017002207.1 | XP_016857696.1 | 3 | 543 | ||
GenBank mRNA | AK225560.1 | - | 3 | 478 | |
AK300399.1 | - | 3 | 407 | ||
AK300623.1 | - | 4 | 429 | ||
BC038239.1 | - | 3 | 502 | 5767075 | |
EU826586.1 | - | 3 | 423 | ||
EU826587.1 | - | 3 | 423 | ||
EU826588.1 | - | 3 | 423 | ||
EU826590.1 | - | 3 | 423 | ||
X60957.1 | - | 3 | 459 |