Gene Transcripts

Gene Symbol
Sprr2d
Entrez Gene ID
20758
Gene Name
small proline-rich protein 2D
Gene Aliases
Chromosome Location
Chr. 3: 92339140 - 92340873 on Build GRCm38
UniGene ID
Mm.87820
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011470.2 NP_035600.1 2 608
GenBank mRNA AJ005562.1 - 2 591
AK008778.1 - 2 609
AY158988.1 - 2 609

View Full Product Details