Gene Transcripts

Gene Symbol
Syk
Entrez Gene ID
20963
Gene Name
spleen tyrosine kinase
Gene Aliases
Sykb
Chromosome Location
Chr. 13: 52582329 - 52648792 on Build GRCm38
UniGene ID
Mm.375031
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001198977.1 NP_001185906.1 11 1603
NM_011518.2 NP_035648.2 11 1607
GenBank mRNA AK049207.1 - 11 1632
AK136363.1 - 11 1606
AK143564.1 - 11 1636
BC065121.1 - 11 1539 30533300
U25685.1 - 10 1391
U36776.1 - 11 1610
Z49877.1 - 11 1865

View Full Product Details