Gene Transcripts

Gene Symbol
Tenm2
Entrez Gene ID
23964
Gene Name
teneurin transmembrane protein 2
Gene Aliases
Odz2, Odz3, Ten-2, Ten-m2
Chromosome Location
Chr. 11: 36006656 - 37236808 on Build GRCm38
UniGene ID
Mm.439889
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290702.1 NP_001277631.1 28 8633
NM_011856.4 NP_035986.3 29 8726
GenBank mRNA AB025411.1 - 29 8764
AF195419.1 - 4 3151
AK122455.1 - 1 4674
AK149173.1 - 1 269

View Full Product Details