Gene Transcripts

Gene Symbol
Slc11a1
Entrez Gene ID
18173
Gene Name
solute carrier family 11 (proton-coupled divalent metal ion transporters), member 1
Gene Aliases
Bcg, Ity, Lsh, Nramp, Nramp1, ity
Chromosome Location
Chr. 1: 74375014 - 74386057 on Build GRCm38
UniGene ID
Mm.2913
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013612.2 NP_038640.2 4 468
GenBank mRNA AK090303.1 - 4 469
AK144237.1 - 4 451
AK171343.1 - 4 397
AK171393.1 - 4 449
BC109137.1 - 4 382 40039390
BC109138.1 - 4 382 40039397
L13732.1 - 4 446
X75355.1 - 4 427

View Full Product Details