Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001161730.1 | NP_001155202.1 | 5 | 1432 | |
NM_013683.2 | NP_038711.2 | 5 | 1432 | ||
GenBank mRNA | AK040629.1 | - | 4 | 1821 | |
AK040919.1 | - | 3 | 2395 | ||
AK151481.1 | - | 5 | 1187 | ||
AK152953.1 | - | 5 | 1132 | ||
AK153890.1 | - | 5 | 1436 | ||
AK162731.1 | - | 5 | 1232 | ||
AK166046.1 | - | 5 | 1433 | ||
AK167485.1 | - | 5 | 1434 | ||
AK171363.1 | - | 5 | 1187 | ||
BC013802.1 | - | 5 | 843 | 3257566 | |
BC024897.1 | - | 5 | 955 | 3257734 | |
BC031584.1 | - | 1 | 1567 | ||
KJ650227.1 | - | 5 | 1108 | ||
M55637.1 | - | 5 | 824 | ||
U60018.1 | - | 5 | 1108 | ||
U60019.1 | - | 5 | 1108 | ||
U60020.1 | - | 5 | 1108 | ||
U60021.1 | - | 5 | 1108 | ||
U60022.1 | - | 5 | 1108 | ||
U60023.1 | - | 5 | 1108 | ||
X59615.1 | - | 5 | 667 |