Gene Transcripts

Gene Symbol
AQP7
Entrez Gene ID
364
Gene Name
aquaporin 7
Gene Aliases
AQP7L, AQPap, GLYCQTL
Chromosome Location
Chr. 9: 33383135 - 33402682 on Build GRCh38
UniGene ID
Hs.455323
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170.2 NP_001161.1 8 1207
NM_001318156.1 NP_001305085.1 7 1030
NM_001318157.1 NP_001305086.1 7 1883
NM_001318158.1 NP_001305087.1 7 1565
NR_134513.1 5 1322
NR_134514.1 6 1441
NR_134515.1 7 1592
XM_005251453.3 XP_005251510.1 9 1307
XM_006716765.3 XP_006716828.1 7 968
XM_011517866.2 XP_011516168.1 8 1117
XM_011517867.2 XP_011516169.1 6 747
XM_017014700.1 XP_016870189.1 8 1408
XM_017014701.1 XP_016870190.1 7 1078
XM_017014702.1 XP_016870191.1 8 1113
XM_017014703.1 XP_016870192.1 7 866
XM_017014704.1 XP_016870193.1 6 1174
XM_017014705.1 XP_016870194.1 6 844
XM_017014706.1 XP_016870195.1 8 1596
GenBank mRNA AB006190.1 - 8 1044
BC062701.1 - 7 857 30324263
BC119672.1 - 6 1273
BC119673.1 - 6 1273
DQ786216.1 - 3 419

View Full Product Details