Gene Transcripts

Gene Symbol
Schip1
Entrez Gene ID
30953
Gene Name
schwannomin interacting protein 1
Gene Aliases
Nf2ip, Schip-1
Chromosome Location
Chr. 3: 68064802 - 68626482 on Build GRCm38
UniGene ID
Mm.331064
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113420.1 NP_001106891.1 7 1206
NM_001113421.1 NP_001106892.1 8 1890
NM_001282044.1 NP_001268973.1 8 1873
NM_001282045.1 NP_001268974.1 8 1731
NM_013928.5 NP_038956.2 7 1296
GenBank mRNA AK021361.1 - 7 1155
AK083464.1 - 8 1715
AK146930.1 - 8 1845
AK168005.1 - 8 1799
BB440451.2 - 1 332
BC060529.1 - 7 1230
BC094348.1 - 8 1657 6414097
BY616163.1 - 1 94
BY623736.1 - 1 55
EU163407.1 - 8 1890
EU163408.1 - 7 1206

Gene Symbol
Gm21949
Entrez Gene ID
100505386
Gene Name
predicted gene, 21949
Gene Aliases
Iqcj-schip1, Schip-1-iqcj, Schip1
Chromosome Location
Chr. 3: 67892220 - 68626482 on Build GRCm38
UniGene ID
Mm.331064
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113419.2 NP_001106890.1 11 2068

View Full Product Details