Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001038710.1 | NP_001033799.1 | 10 | 2313 | |
NM_016711.3 | NP_057920.2 | 10 | 2249 | ||
GenBank mRNA | AK046844.1 | - | 1 | 1519 | |
AK079077.1 | - | 10 | 2094 | ||
AK147581.1 | - | 10 | 2246 | ||
AK164178.1 | - | 10 | 2096 |