Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016843.3 | NP_058539.2 | 5 | 658 | |
GenBank mRNA | AK002504.1 | - | 5 | 767 | |
AK003530.1 | - | 5 | 585 | ||
AK047170.1 | - | 5 | 708 | ||
AK049706.1 | - | 5 | 702 | ||
AK077738.1 | - | 5 | 660 | ||
AK078459.1 | - | 5 | 738 | ||
AK153271.1 | - | 5 | 660 | ||
AK161950.1 | - | 675 | |||
AK165941.1 | - | 5 | 683 | ||
AK166899.1 | - | 5 | 743 | ||
AK170507.1 | - | 5 | 675 | ||
AK170751.1 | - | 5 | 684 | ||
AK210764.1 | - | 2 | 152 | ||
BC016410.1 | - | 5 | 668 | 3968016 | |
BC046802.1 | - | 5 | 794 | 6404607 | |
X61506.1 | - | 5 | 772 |