Gene Transcripts

Gene Symbol
Atxn10
Entrez Gene ID
54138
Gene Name
ataxin 10
Gene Aliases
AI325283, C77170, E46, Sca10, TEG-169, Tex169
Chromosome Location
Chr. 15: 85336381 - 85463836 on Build GRCm38
UniGene ID
Mm.248906
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016843.3 NP_058539.2 7 878
GenBank mRNA AK002504.1 - 7 987
AK003530.1 - 7 805
AK047170.1 - 7 928
AK049706.1 - 7 922
AK077738.1 - 7 881
AK078459.1 - 7 958
AK153271.1 - 7 880
AK161950.1 - 895
AK165941.1 - 7 903
AK166899.1 - 7 963
AK170507.1 - 7 895
AK170751.1 - 7 904
AK195052.1 - 2 112
AK219896.1 - 2 112
BC002285.1 - 1 47
BC016410.1 - 7 888 3968016
BC046802.1 - 7 1014 6404607
X61506.1 - 7 992

View Full Product Details