Gene Transcripts

Gene Symbol
Gosr2
Entrez Gene ID
56494
Gene Name
golgi SNAP receptor complex member 2
Gene Aliases
2310032N09Rik, C76855, Gs27, SNARE, membrin
Chromosome Location
Chr. 11: 103676849 - 103697738 on Build GRCm38
UniGene ID
Mm.195451
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019650.3 NP_062624.2 3 231
GenBank mRNA AF007550.1 - 3 179
AK002556.1 - 3 221
AK005697.1 - 3 209
AK013052.1 - 3 225
AK150004.1 - 3 229
BC008525.1 - 3 227 3601627
BC051253.1 - 3 197 6485874

View Full Product Details