Gene Transcripts

Gene Symbol
Fam60a
Entrez Gene ID
56306
Gene Name
family with sequence similarity 60, member A
Gene Aliases
Ppcs1, Pptcs1, Tera
Chromosome Location
Chr. 6: 148921045 - 148946432 on Build GRCm38
UniGene ID
Mm.18637
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019643.3 NP_062617.2 3 373
GenBank mRNA AK046519.1 - 3 280
AK145384.1 - 3 384
AK166569.1 - 3 379
BC076599.1 - 3 373 30614283
U64033.1 - 3 462

View Full Product Details