Gene Transcripts
- Gene Symbol
- Ncoa4
- Entrez Gene ID
- 27057
- Gene Name
- nuclear receptor coactivator 4
- Gene Aliases
- ARA70, Rfg
- Chromosome Location
- Chr. 14: 32159887 - 32179855 on Build GRCm38
- UniGene ID
- Mm.371598
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001033988.2 |
NP_001029160.1
|
8, 9 |
1877 |
|
NM_001284319.1 |
NP_001271248.1
|
8, 9 |
1869 |
|
NM_019744.4 |
NP_062718.2
|
8, 9 |
1787 |
|
GenBank mRNA |
AF159461.1 |
- |
8, 9 |
1784 |
|
AK014479.1 |
- |
8, 9 |
1788 |
|
AK016355.1 |
- |
1, 2 |
55 |
|
AK032951.1 |
- |
6, 7 |
1543 |
|
AK129020.1 |
- |
8, 9 |
1835 |
|
AK143965.1 |
- |
8, 9 |
1781 |
|
AK145945.1 |
- |
8, 9 |
1830 |
|
AK145954.1 |
- |
8, 9 |
1834 |
|
AK146384.1 |
- |
8, 9 |
1788 |
|
AK146498.1 |
- |
8, 9 |
1827 |
|
AK146598.1 |
- |
8, 9 |
1811 |
|
AK146646.1 |
- |
8, 9 |
1782 |
|
AK146649.1 |
- |
8, 9 |
1784 |
|
AK147036.1 |
- |
8, 9 |
1786 |
|
AK147125.1 |
- |
8, 9 |
1829 |
|
AK156999.1 |
- |
8, 9 |
1869 |
|
BC085086.1 |
- |
8, 9 |
1773 |
6335192 |
View Full Product Details