Gene Transcripts

Gene Symbol
Amotl2
Entrez Gene ID
56332
Gene Name
angiomotin-like 2
Gene Aliases
AW549739, C79691, Lccp, MASCOT, mKIAA0989
Chromosome Location
Chr. 9: 102717294 - 102733418 on Build GRCm38
UniGene ID
Mm.21145
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019764.2 NP_062738.2 10 3386
GenBank mRNA AF175967.1 - 10 2669
AF175968.1 - 8 3526
AK040912.1 - 10 3467
AK077535.1 - 10 3384
AK141348.1 - 10 3386
AK164279.1 - 10 3455
AK164819.1 - 11 3489
AK169419.1 - 10 3386
BC027824.1 - 10 3356 5323888
BC035201.1 - 10 3368
BC108411.1 - 10 3420

View Full Product Details