Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001195007.1 | NP_001181936.1 | 8 | 604 | |
NM_001319293.1 | NP_001306222.1 | 8 | 565 | ||
NM_006112.3 | NP_006103.1 | 8 | 604 | ||
NM_203456.2 | NP_982281.1 | 8 | 604 | ||
NR_036543.1 | 8 | 596 | |||
NR_036544.1 | 8 | 557 | |||
XM_006710289.2 | XP_006710352.1 | 8 | 520 | ||
XM_006710290.3 | XP_006710353.1 | 8 | 520 | ||
XM_011540501.1 | XP_011538803.1 | 8 | 706 | ||
XM_017000051.1 | XP_016855540.1 | 8 | 555 | ||
XM_017000052.1 | XP_016855541.1 | 8 | 706 | ||
GenBank mRNA | AF042385.1 | - | 8 | 608 | |
AF042386.1 | - | 8 | 608 | ||
AF104012.1 | - | 8 | 559 | ||
AF104013.1 | - | 8 | 559 | ||
AK094660.1 | - | 7 | 1831 | ||
AK130628.1 | - | 8 | 557 | ||
AK293094.1 | - | 8 | 555 | ||
AK304694.1 | - | 8 | 512 | ||
AK313666.1 | - | 8 | 572 | ||
AL833576.1 | - | 8 | 3931 | ||
BC004898.2 | - | 8 | 554 | 3545997 | |
BC008451.1 | - | 8 | 567 | 4134572 | |
BC107736.1 | - | 8 | 563 | 4809032 | |
BU160972.1 | - | 8 | 533 | ||
BU179717.1 | - | 591 | |||
BU196539.1 | - | 8 | 591 | ||
CT002466.1 | - | 8 | 506 | ||
DQ160195.1 | - | 8 | 548 |