Gene Transcripts

Gene Symbol
Ckb
Entrez Gene ID
12709
Gene Name
creatine kinase, brain
Gene Aliases
B-CK, Bck, Ck-3, Ck3, Ckbb
Chromosome Location
Chr. 12: 111669355 - 111672338 on Build GRCm38
UniGene ID
Mm.16831
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021273.4 NP_067248.1 3 369
GenBank mRNA AK002467.1 - 3 332
AK014299.1 - 3 312
AK148885.1 - 3 329
AK152388.1 - 3 329
AK153484.1 - 3 329
AK161990.1 - 3 329
AK165779.1 - 1 777
AK166980.1 - 3 332
AK167034.1 - 3 332
BC015271.1 - 3 299 4225384
BC058257.1 - 1 755
BC106109.1 - 3 296 6395794

View Full Product Details