Gene Transcripts

Gene Symbol
Prokr1
Entrez Gene ID
58182
Gene Name
prokineticin receptor 1
Gene Aliases
EG-VEGFR1, Gpr73, Pkr1
Chromosome Location
Chr. 6: 87578589 - 87590774 on Build GRCm38
UniGene ID
Mm.333226
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021381.3 NP_067356.2 2 401
GenBank mRNA AF236082.1 - 1 155
AF487278.1 - 1 155
AK134279.1 - 2 415
AK156734.1 - 2 575
BC059003.1 - 2 401 6848525

View Full Product Details