Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021435.3 | NP_067410.1 | 5 | 596 | |
GenBank mRNA | AB041549.1 | - | 5 | 597 | |
AK029046.1 | - | 4 | 3217 | ||
AK029969.1 | - | 5 | 654 | ||
AK033913.1 | - | 5 | 526 | ||
AK042150.1 | - | 5 | 496 | ||
AK045791.1 | - | 5 | 539 | ||
AK138591.1 | - | 5 | 537 | ||
BC016275.1 | - | 5 | 527 | ||
BC033512.1 | - | 5 | 521 | 4017336 |