Gene Transcripts

Gene Symbol
Pcdh15
Entrez Gene ID
11994
Gene Name
protocadherin 15
Gene Aliases
BB078305, ENSMUSG00000046980, Gm9815, Ush1f, av, nmf19
Chromosome Location
Chr. 10: 73100028 - 74649737 on Build GRCm38
UniGene ID
Mm.338933
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142735.1 NP_001136207.1 4 663
NM_001142736.1 NP_001136208.1 4 663
NM_001142737.1 NP_001136209.1 4 663
NM_001142738.1 NP_001136210.1 4 663
NM_001142739.1 NP_001136211.1 3 597
NM_001142740.1 NP_001136212.1 4 663
NM_001142741.1 NP_001136213.1 3 597
NM_001142742.1 NP_001136214.1 5 678
NM_001142743.1 NP_001136215.1 4 663
NM_001142746.1 NP_001136218.1 5 678
NM_001142747.1 NP_001136219.1 5 678
NM_001142748.1 NP_001136220.1 4 663
NM_001142760.1 NP_001136232.1 4 663
NM_023115.3 NP_075604.2 5 678
GenBank mRNA AF281899.1 - 5 662
AK139154.1 - 5 603
DQ354396.1 - 4 647
DQ354397.1 - 4 647
DQ354398.1 - 4 647
DQ354399.1 - 4 647
DQ354400.1 - 4 647
DQ354401.1 - 4 647
DQ354402.1 - 3 581
DQ354403.1 - 4 647
DQ354404.1 - 3 581
DQ354405.1 - 5 662
DQ354406.1 - 4 647
DQ354408.1 - 4 647
DQ354409.1 - 4 647
DQ354410.1 - 4 647
DQ354411.1 - 4 647
DQ354412.1 - 4 647
DQ354413.1 - 5 662
DQ354414.1 - 4 647
DQ354416.1 - 5 603
DQ354417.1 - 4 588
DQ354418.1 - 4 588

View Full Product Details