Gene Transcripts

Gene Symbol
AQP9
Entrez Gene ID
366
Gene Name
aquaporin 9
Gene Aliases
AQP-9, HsT17287, SSC1, T17287
Chromosome Location
Chr. 15: 58138169 - 58185911 on Build GRCh38
UniGene ID
Hs.104624
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320635.1 NP_001307564.1 5 2369
NM_001320636.1 NP_001307565.1 6 2219
NM_020980.4 NP_066190.2 6 2587
GenBank mRNA AB008775.1 - 6 2433
AF016495.1 - 6 2444
BC026258.1 - 7 2356 4734559

View Full Product Details