Gene Transcripts
- Gene Symbol
- C8B
- Entrez Gene ID
- 732
- Gene Name
- complement component 8, beta polypeptide
- Gene Aliases
- C82
- Chromosome Location
- Chr. 1: 56929210 - 56974383 on Build GRCh38
- UniGene ID
- Hs.391835
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000066.3 |
NP_000057.2
|
1, 2 |
294 |
|
NM_001278543.1 |
NP_001265472.1
|
2, 3 |
516 |
|
NM_001278544.1 |
NP_001265473.1
|
1, 2 |
294 |
|
XM_017002235.1 |
XP_016857724.1
|
2, 3 |
362 |
|
GenBank mRNA |
AK292740.1 |
- |
1, 2 |
149 |
|
AK298425.1 |
- |
1, 2 |
211 |
|
AK298444.1 |
- |
2, 3 |
516 |
|
AK313382.1 |
- |
1, 2 |
166 |
|
AK316209.1 |
- |
2, 3 |
513 |
|
BC020701.1 |
- |
1, 2 |
169 |
|
BC130575.1 |
- |
1, 2 |
127 |
40146606 |
M16973.1 |
- |
1, 2 |
129 |
|
View Full Product Details