Gene Transcripts

Gene Symbol
Psmd6
Entrez Gene ID
66413
Gene Name
proteasome (prosome, macropain) 26S subunit, non-ATPase, 6
Gene Aliases
2400006A19Rik
Chromosome Location
Chr. 14: 14112185 - 14120904 on Build GRCm38
UniGene ID
Mm.27591
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025550.2 NP_079826.2 2 296
GenBank mRNA AJ252147.1 - 2 332
AK002343.1 - 2 330
AK010287.1 - 2 329
AK211367.1 - 1 29
BC006869.1 - 2 296 2646325

View Full Product Details