Gene Transcripts

Gene Symbol
Cyc1
Entrez Gene ID
66445
Gene Name
cytochrome c-1
Gene Aliases
2610002H19Rik, AA408921
Chromosome Location
Chr. 15: 76343523 - 76345934 on Build GRCm38
UniGene ID
Mm.29196
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025567.2 NP_079843.1 4 554
GenBank mRNA AK002815.1 - 4 532
AK011288.1 - 4 555
AK151299.1 - 4 545
AK170272.1 - 3 1154
AK186904.1 - 2 117
AK192516.1 - 4 411
BC005620.1 - 4 535 3711323
BC082790.1 - 4 492

View Full Product Details