Gene Transcripts
- Gene Symbol
- Rpe
- Entrez Gene ID
- 66646
- Gene Name
- ribulose-5-phosphate-3-epimerase
- Gene Aliases
- 2810429B02Rik, 5730518J08Rik
- Chromosome Location
- Chr. 1: 66700797 - 66719805 on Build GRCm38
- UniGene ID
- Mm.240912
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_025683.2 |
NP_079959.2
|
2, 3 |
211 |
|
GenBank mRNA |
AK007735.1 |
- |
2, 3 |
217 |
|
AK017774.1 |
- |
2, 3 |
212 |
|
AK029088.1 |
- |
2, 3 |
210 |
|
AK088152.1 |
- |
2, 3 |
212 |
|
AK150008.1 |
- |
2, 3 |
249 |
|
AK185217.1 |
- |
2, 3 |
205 |
|
BC006953.1 |
- |
2, 3 |
205 |
|
BC019126.1 |
- |
2, 3 |
254 |
5008075 |
Z78161.1 |
- |
3, 4 |
230 |
|
View Full Product Details