Gene Transcripts

Gene Symbol
Sike1
Entrez Gene ID
66641
Gene Name
suppressor of IKBKE 1
Gene Aliases
2810005O12Rik, 5730470L24Rik, AI450236, AI839862, Sike, Sikeb
Chromosome Location
Chr. 3: 102995740 - 103003914 on Build GRCm38
UniGene ID
Mm.208619
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025679.3 NP_079955.1 5 1238
GenBank mRNA AK017694.1 - 5 1234
AK033553.1 - 5 1242
AK035590.1 - 5 1252

View Full Product Details