Gene Transcripts
- Gene Symbol
- F8
- Entrez Gene ID
- 2157
- Gene Name
- coagulation factor VIII
- Gene Aliases
- AHF, DXS1253E, F8B, F8C, FVIII, HEMA
- Chromosome Location
- Chr. X: 154835788 - 155022723 on Build GRCh38
- UniGene ID
- Hs.632836
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000132.3 |
NP_000123.1
|
24, 25 |
6902 |
|
NM_019863.2 |
NP_063916.1
|
3, 4 |
471 |
|
GenBank mRNA |
AK289947.1 |
- |
3, 4 |
401 |
|
AK295164.1 |
- |
12, 13 |
1699 |
|
AK313707.1 |
- |
3, 4 |
356 |
|
AX746360.1 |
- |
24, 25 |
6731 |
|
BC022513.1 |
- |
3, 4 |
390 |
|
BC064380.1 |
- |
3, 4 |
450 |
|
BC098389.1 |
- |
3, 4 |
469 |
|
BC111967.1 |
- |
3, 4 |
359 |
|
BC111969.1 |
- |
3, 4 |
358 |
|
K01740.1 |
- |
24, 25 |
6881 |
|
KT878539.1 |
- |
1, 2 |
152 |
|
M14113.1 |
- |
24, 25 |
6902 |
|
M90707.1 |
- |
3, 4 |
366 |
|
X01179.1 |
- |
24, 25 |
6840 |
|
View Full Product Details