Gene Transcripts

Gene Symbol
Psmd12
Entrez Gene ID
66997
Gene Name
proteasome (prosome, macropain) 26S subunit, non-ATPase, 12
Gene Aliases
1500002F15Rik, AI480719, P55
Chromosome Location
Chr. 11: 107479528 - 107498036 on Build GRCm38
UniGene ID
Mm.491171
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025894.2 NP_080170.1 1, 2 165
GenBank mRNA AK005114.1 - 1, 2 200
AK007619.1 - 1, 2 210
AK007680.1 - 1, 2 139
AK008646.1 - 1, 2 166
AK009969.1 - 1, 2 168
AK011806.1 - 1, 2 189
AK017843.1 - 1, 2 182
AK150910.1 - 1, 2 189
AK151242.1 - 1, 2 166
AK152041.1 - 1, 2 189
AK152240.1 - 1, 2 197
AK160283.1 - 1, 2 187
AK162779.1 - 166
AK167236.1 - 1, 2 173
BC004694.1 - 1, 2 183 3498952

View Full Product Details