Gene Transcripts

Gene Symbol
FUT6
Entrez Gene ID
2528
Gene Name
fucosyltransferase 6
Gene Aliases
FCT3A, FT1A, Fuc-TVI, FucT-VI
Chromosome Location
Chr. 19: 5830626 - 5839753 on Build GRCh38
UniGene ID
Hs.631846
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000150.2 NP_000141.1 3 2311
NM_001040701.1 NP_001035791.1 2 2183
XM_005259526.4 XP_005259583.1 4 2378
XM_011527868.2 XP_011526170.1 4 2450
XM_011527869.2 XP_011526171.1 4 2447
XM_011527870.2 XP_011526172.1 5 2558
XM_011527872.2 XP_011526174.1 5 2578
XM_011527873.2 XP_011526175.1 4 2513
XM_011527874.2 XP_011526176.1 4 2533
XM_011527875.2 XP_011526177.1 5 2604
XM_011527876.2 XP_011526178.1 4 2559
XM_011527878.2 XP_011526180.1 4 2450
XM_011527879.2 XP_011526181.1 3 2405
GenBank mRNA AF131211.1 - 4 1392
AK098073.1 - 3 1168
AK225795.1 - 2 1690
BC061700.1 - 2 2173 4606746
U27333.1 - 3 2269
U27334.1 - 3 1592
U27335.1 - 2 2144
U27336.1 - 2 1467
U27337.1 - 3 1754

View Full Product Details