Gene Transcripts

Gene Symbol
Zmynd19
Entrez Gene ID
67187
Gene Name
zinc finger, MYND domain containing 19
Gene Aliases
2700064H14Rik, AA536891, AU014915, mizip
Chromosome Location
Chr. 2: 24949802 - 24960200 on Build GRCm38
UniGene ID
Mm.296106
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026021.3 NP_080297.1 2, 3 314
GenBank mRNA AJ409150.1 - 2, 3 275
AJ409151.1 - 2, 3 160
AK005908.1 - 2, 3 302
AK012484.1 - 2, 3 315
AK088063.1 - 2, 3 336
BC027321.1 - 2, 3 280 3990966

View Full Product Details