Gene Transcripts

Gene Symbol
Aasdhppt
Entrez Gene ID
67618
Gene Name
aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase
Gene Aliases
2010309J24Rik, 2810407B07Rik, AASD-PPT, CGI-80, LYS2, LYS5
Chromosome Location
Chr. 9: 4294793 - 4309509 on Build GRCm38
UniGene ID
Mm.33970
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026276.3 NP_080552.3 2 441
GenBank mRNA AK007737.1 - 2 442
AK010557.1 - 2 401
AK013111.1 - 2 395
AK014188.1 - 2 384
AK018191.1 - 1 87
AK028700.1 - 2 398
AK029523.1 - 2 383
AK031766.1 - 2 385
AK076099.1 - 2 396
AK076866.1 - 2 415
AK081818.1 - 2 417
AK088977.1 - 2 381
AK144671.1 - 2 415
AK214035.1 - 1 109
AK215948.1 - 1 109
BC030043.1 - 2 376 1348651
BC038013.1 - 2 412 3497754
BC049851.1 - 2 402 3481972

View Full Product Details