Gene Transcripts

Gene Symbol
Lce1f
Entrez Gene ID
67828
Gene Name
late cornified envelope 1F
Gene Aliases
1110055J05Rik
Chromosome Location
Chr. 3: 92718696 - 92720350 on Build GRCm38
UniGene ID
Mm.158176
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026394.3 NP_080670.2 2 667
GenBank mRNA AK003518.1 - 2 668
BC060283.1 - 2 651
BC100541.1 - 1 635

View Full Product Details