Gene Transcripts

Gene Symbol
Atad1
Entrez Gene ID
67979
Gene Name
ATPase family, AAA domain containing 1
Gene Aliases
4921525H23Rik, AW107648, Thorase
Chromosome Location
Chr. 19: 32672558 - 32739766 on Build GRCm38
UniGene ID
Mm.27123
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026487.3 NP_080763.2 6, 7 744
GenBank mRNA AK007147.1 - 2, 3 201
AK009419.1 - 2, 3 332
AK014967.1 - 6, 7 761
AK030719.1 - 6, 7 764
AK033639.1 - 6, 7 768
AK044925.1 - 6, 7 744
AK150469.1 - 6, 7 764
AK152059.1 - 6, 7 764
AK165953.1 - 6, 7 779
BC029085.1 - 6, 7 744 3593451
BC043051.1 - 6, 7 729 5708377

View Full Product Details