Gene Transcripts

Gene Symbol
Rps13
Entrez Gene ID
68052
Gene Name
ribosomal protein S13
Gene Aliases
2700063M04Rik
Chromosome Location
Chr. 7: 116331507 - 116334190 on Build GRCm38
UniGene ID
Mm.14798
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026533.3 NP_080809.1 4 349
GenBank mRNA AK012479.1 - 4 294
AK018703.1 - 4 293
AK076060.1 - 4 293
AK088237.1 - 3 926
AK151043.1 - 4 294
AK151671.1 - 4 295
AK161798.1 - 3 892
AK166691.1 - 4 294
AK177445.1 - 292
AV498110.1 - 3 256
BC011192.1 - 4 292 4016679
BC085269.1 - 2 998
BC090397.1 - 4 276 5150054
BC106137.1 - 4 277 4990874
BC125602.1 - 4 293 40130117
BC125604.1 - 4 293 40130119

View Full Product Details