Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026558.4 | NP_080834.3 | 2, 3 | 555 | |
GenBank mRNA | AK013129.1 | - | 2, 3 | 396 | |
AK019574.1 | - | 2, 3 | 383 | ||
AK078438.1 | - | 2, 3 | 536 | ||
AK088705.1 | - | 2, 3 | 536 | ||
BC020162.1 | - | 349 | 3991990 |