Gene Transcripts

Gene Symbol
Mcoln2
Entrez Gene ID
68279
Gene Name
mucolipin 2
Gene Aliases
3300002C04Rik, AI549968, C86638, TRPML2
Chromosome Location
Chr. 3: 146149796 - 146195514 on Build GRCm38
UniGene ID
Mm.116862
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005846.2 NP_001005846.1 8, 9 1034
NM_026656.4 NP_080932.2 8, 9 1062
GenBank mRNA AF503575.1 - 8, 9 1051
AK014467.1 - 8, 9 965
AK019454.1 - 8, 9 931
AK150446.1 - 8, 9 1030
AK150547.1 - 8, 9 1030
AK152067.1 - 8, 9 1030
AK152943.1 - 8, 9 1030
AK153290.1 - 8, 9 1030
AK159172.1 - 8, 9 1062
AY083532.1 - 7, 8 880
BC029847.1 - 8, 9 911 3967456

View Full Product Details