Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001291050.1 | NP_001277979.1 | 19 | 2428 | |
NM_001291051.1 | NP_001277980.1 | 19 | 2458 | ||
NM_001291052.1 | NP_001277981.1 | 19 | 2476 | ||
NM_027096.2 | NP_081372.1 | 18 | 2352 | ||
NM_028802.3 | NP_083078.3 | 20 | 2552 | ||
GenBank mRNA | AK009563.1 | - | 13 | 1771 | |
AK030645.1 | - | 20 | 2506 | ||
AK031658.1 | - | 19 | 2431 | ||
AK050318.1 | - | 20 | 2467 | ||
AK051728.1 | - | 12 | 1615 | ||
AK122510.1 | - | 7 | 3207 | ||
AK165952.1 | - | 20 | 2519 | ||
AK166293.1 | - | 11 | 1486 | ||
AK166454.1 | - | 19 | 2440 | ||
BC006887.1 | - | 19 | 2400 | ||
BC033408.1 | - | 18 | 2282 | 5357683 |