Gene Transcripts

Gene Symbol
Spsb3
Entrez Gene ID
79043
Gene Name
splA/ryanodine receptor domain and SOCS box containing 3
Gene Aliases
2310012N15Rik, 3300001M01Rik, SSB3, Tce1, mKIAA4204
Chromosome Location
Chr. 17: 24886349 - 24892147 on Build GRCm38
UniGene ID
Mm.274794
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163750.1 NP_001157222.1 3 651
NM_001163751.1 NP_001157223.1 3 306
NM_027141.2 NP_081417.2 3 327
GenBank mRNA AF403038.1 - 3 327
AK009325.1 - 3 327
AK076170.1 - 3 603
AK082768.1 - 3 250
AK156579.1 - 3 589
AK167474.1 - 3 567
AK199452.1 - 3 220
AK218464.1 - 2 135
AK220234.1 - 3 589
BC026523.1 - 3 992
BC072556.1 - 3 612 5691114
BY276751.1 - 3 307

View Full Product Details