Gene Transcripts

Gene Symbol
Rasip1
Entrez Gene ID
69903
Gene Name
Ras interacting protein 1
Gene Aliases
1110025D03Rik, 2610025P08Rik, AI853551, Rain
Chromosome Location
Chr. 7: 45627537 - 45639092 on Build GRCm38
UniGene ID
Mm.181396
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028544.1 NP_082820.1 10, 11 2604
GenBank mRNA AK011557.2 - 4, 5 687
AK033276.1 - 8, 9 1892
AK156602.1 - 10, 11 2616
BC072584.1 - 7, 8 1558
BC127059.1 - 8, 9 1530 40109852
BC150766.1 - 10, 11 2604 9087677

View Full Product Details