Gene Transcripts

Gene Symbol
Wdr20rt
Entrez Gene ID
70948
Gene Name
WD repeat domain 20, retrogene
Gene Aliases
4921538B03Rik, 4930427E19Rik, Wdr20, Wdr20b
Chromosome Location
Chr. 12: 65225517 - 65228454 on Build GRCm38
UniGene ID
Mm.219475
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027614.1 NP_081890.1 1 537
GenBank mRNA AK015014.1 - 1 537
BC050778.1 - 1 324 6743775

View Full Product Details