Gene Transcripts

Gene Symbol
Pdgfd
Entrez Gene ID
71785
Gene Name
platelet-derived growth factor, D polypeptide
Gene Aliases
1110003I09Rik
Chromosome Location
Chr. 9: 6168362 - 6536014 on Build GRCm38
UniGene ID
Mm.390122
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027924.2 NP_082200.1 2 350
GenBank mRNA AF335583.1 - 2 167
AK003359.2 - 2 352
AK141551.1 - 2 361
BC030896.1 - 2 301 4489485

View Full Product Details