Gene Transcripts

Gene Symbol
Dbndd1
Entrez Gene ID
72185
Gene Name
dysbindin (dystrobrevin binding protein 1) domain containing 1
Gene Aliases
2810427I04Rik, D8Ertd590e
Chromosome Location
Chr. 8: 123504718 - 123518041 on Build GRCm38
UniGene ID
Mm.27925
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170975.2 NP_001164446.2 2 332
NM_001170976.1 NP_001164447.2 2 332
NM_028146.4 NP_082422.3 2 332
GenBank mRNA AK013173.1 - 2 231
AK028187.1 - 2 269
AK163434.1 - 2 332
BC046622.1 - 2 267 6389902
BQ554936.1 - 2 239

View Full Product Details