Gene Transcripts

Gene Symbol
Mfsd8
Entrez Gene ID
72175
Gene Name
major facilitator superfamily domain containing 8
Gene Aliases
2810423E13Rik, AI836898, AV142426, Cln7
Chromosome Location
Chr. 3: 40818112 - 40846856 on Build GRCm38
UniGene ID
Mm.130610
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028140.4 NP_082416.2 8 944
GenBank mRNA AK013144.1 - 8 836
AK032080.1 - 8 965
AK035291.1 - 8 908
AK077957.1 - 8 945
BC010483.1 - 8 793
BC113183.1 - 8 875 40061891
BC113790.1 - 8 875 40061889

View Full Product Details