Gene Transcripts

Gene Symbol
TGFBI
Entrez Gene ID
7045
Gene Name
transforming growth factor beta induced
Gene Aliases
BIGH3, CDB1, CDG2, CDGG1, CSD, CSD1, CSD2, CSD3, EBMD, LCD1
Chromosome Location
Chr. 5: 136028895 - 136063818 on Build GRCh38
UniGene ID
Hs.369397
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000358.2 NP_000349.1 2 328
GenBank mRNA AK093916.1 - 2 183
AK094055.1 - 3 198
AK094581.1 - 2 272
AK222833.1 - 2 236
AK303643.1 - 2 232
BC000097.2 - 2 221 3506051
BC004972.1 - 2 216 2958878
BT009820.1 - 2 167
DA051240.1 - 2 328
KU178357.1 - 2 167
KU178358.1 - 2 167
M77349.1 - 2 214

View Full Product Details