Gene Transcripts
- Gene Symbol
- Ifitm7
- Entrez Gene ID
- 74482
- Gene Name
- interferon induced transmembrane protein 7
- Gene Aliases
- 4933438K12Rik, Mil4
- Chromosome Location
- Chr. 16: 13981479 - 13986867 on Build GRCm38
- UniGene ID
- Mm.484026
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK017110.1 |
- |
|
1864 |
|
View Full Product Details